5-Bromo-2-methylimidazo[1,2-a]pyridine ≥97%
Supplier: Matrix Scientific
069233-1G
069233-5G
101939-398EA
2692.31
USD
101939-398
101939-396
5-Bromo-2-methylimidazo[1,2-a]pyridine ≥97%
5-Bromo-2-methylimidazo[1,2-a]pyridine
Hazard: Irritant
Formula:
C₈H₇BrN₂ MW: 211.0600 g/mol |
MDL Number:
MFCD10699211 CAS Number: 74420-51-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...