6-Bromo-2-chloromethylimidazo[1,2-a]pyridine ≥96%
Supplier: Matrix Scientific
069232-1G
069232-5G
101939-394EA
3790.77
USD
101939-394
101939-392
6-Bromo-2-chloromethylimidazo[1,2-a]pyridine ≥96%
6-Bromo-2-chloromethylimidazo[1,2-a]pyridine
Hazard: Irritant
Formula:
C₈H₆BrClN₂ MW: 245.51 g/mol |
MDL Number:
MFCD06739243 CAS Number: 136117-72-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...