5-Bromo-3-methylimidazo[1,2-a]pyridine ≥97%

Supplier: Matrix Scientific

069230-5G 069230-1G
101939-384EA 1803.08 USD
101939-384 101939-386
5-Bromo-3-methylimidazo[1,2-a]pyridine ≥97%
5-Bromo-3-methylimidazo[1,2-a]pyridine

Hazard: Irritant

Formula: C₈H₇BrN₂
MW: 211.06 g/mol
MDL Number: MFCD10699210
CAS Number: 4926-54-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR