3-Fluorooxetane ≥95%
Supplier: Matrix Scientific
069208-250MG
069208-500MG
101939-270EA
1623.08
USD
101939-270
101939-268
3-Fluorooxetane ≥95%
3-Fluorooxetane
Hazard: Irritant
Formula:
C₃H₅FO |
MDL Number:
MFCD17170023 CAS Number: 26272-86-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...