1,1,1-Trifluoro-2-[(2-methoxyethoxy)methoxy]ethane ≥96%

Supplier: Matrix Scientific

069185-1G 069185-25G 069185-5G
101939-230EA 604.62 USD
101939-230 101939-228 101939-232
1,1,1-Trifluoro-2-[(2-methoxyethoxy)methoxy]ethane ≥96%
1,1,1-Trifluoro-2-[(2-methoxyethoxy)methoxy]ethane

Hazard: Irritant

Formula: C₆H₁₁F₃O₃
MDL Number: MFCD18325133
CAS Number: 130156-55-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR