tert-Butyl-7,7-dimethyl-6,7-dihydro-1H-pyrazolo[4,3-c]pyridine-5(4H)-carboxylate ≥95%

Supplier: Matrix Scientific

069159-1G 069159-5G 069159-500MG
101939-130EA 515.38 USD
101939-130 101939-134 101939-132
tert-Butyl-7,7-dimethyl-6,7-dihydro-1H-pyrazolo[4,3-c]pyridine-5(4H)-carboxylate ≥95%
tert-Butyl-7,7-dimethyl-6,7-dihydro-1H-pyrazolo[4,3-c]pyridine-5(4H)-carboxylate

Hazard: Irritant

Formula: C₁₃H₂₁N₃O₂
MW: 251.33 g/mol
MDL Number: MFCD09878669
CAS Number: 635712-88-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR