5-(Chloromethyl)-1,3-dimethyl-1H-pyrazole ≥98%

Supplier: Matrix Scientific

069154-1G 069154-5G
101939-108EA 276.92 USD
101939-108 101939-110
5-(Chloromethyl)-1,3-dimethyl-1H-pyrazole ≥98%
5-(Chloromethyl)-1,3-dimethyl-1H-pyrazole

Hazard: Irritant

Formula: C₆H₉ClN₂
MW: 144.61 g/mol
MDL Number: MFCD07368502
CAS Number: 852227-86-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR