5-Chloroisoquinoline ≥97%

Supplier: Matrix Scientific

069152-5G
101939-102EA 3547.69 USD
101939-102
5-Chloroisoquinoline ≥97%
5-Chloroisoquinoline

Hazard: Irritant

Formula: C₉H₆ClN
MW: 163.6 g/mol
MDL Number: MFCD02683295
CAS Number: 5430-45-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR