Isoquinoline-8-amine ≥97%
Supplier: Matrix Scientific
Synonyms:
Isoquinolin-8-amine
, 8-Aminoisoquinoline
069151-5G
101939-100EA
267.69
USD
101939-100
Isoquinoline-8-amine ≥97%
Isoquinoline-8-amine
Hazard: Irritant
Formula:
C₉H₈N₂ MW: 144.1700 g/mol |
MDL Number:
MFCD00179553 CAS Number: 23687-27-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...