Dimethyl-1-benzoyl-5-oxopiperidine-2,4-dicarboxylate ≥95%

Supplier: Matrix Scientific

069137-250MG 069137-500MG
101939-054EA 524.62 USD
101939-054 101939-056
Dimethyl-1-benzoyl-5-oxopiperidine-2,4-dicarboxylate ≥95%
Dimethyl-1-benzoyl-5-oxopiperidine-2,4-dicarboxylate

Hazard: Irritant

Formula: C₁₆H₁₇NO₆
MDL Number: MFCD18433658
CAS Number: 1255663-98-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR