(E)-3-(3,4-Dihydroxyphenyl)-1-(2-hydroxy-4,5-dimethylphenyl)prop-2-en-1-one ≥95%
Supplier: Matrix Scientific
069132-500MG
069132-1G
101939-048EA
5521.54
USD
101939-048
101939-046
(E)-3-(3,4-Dihydroxyphenyl)-1-(2-hydroxy-4,5-dimethylphenyl)prop-2-en-1-one ≥95%
(E)-3-(3,4-Dihydroxyphenyl)-1-(2-hydroxy-4,5-dimethylphenyl)prop-2-en-1-one
Hazard: Irritant
Formula:
C₁₇H₁₆O₄ |
MDL Number:
MFCD19443934 CAS Number: 1313738-88-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...