tert-Butyl-4-(cyclopropylmethyl)piperidine-1-carboxylate ≥95%

Supplier: Matrix Scientific

069121-250MG 069121-500MG
101939-018EA 370.77 USD
101939-018 101939-016
tert-Butyl-4-(cyclopropylmethyl)piperidine-1-carboxylate ≥95%
tert-Butyl-4-(cyclopropylmethyl)piperidine-1-carboxylate

Hazard: Irritant

Formula: C₁₄H₂₅NO₂
MW: 239.36 g/mol
MDL Number: MFCD17015066
CAS Number: 1241725-67-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR