tert-Butyl-9-oxo-4,8-diazaspiro[4.4]nonane-4-carboxylate ≥95%

Supplier: Matrix Scientific

Synonyms: tert-Butyl 6-oxo-1,7-diazaspiro[4.4]nonane-1-carboxylate

069114-250MG 069114-500MG
101938-986EA 524.62 USD
101938-986 101938-988
tert-Butyl-9-oxo-4,8-diazaspiro[4.4]nonane-4-carboxylate ≥95%
tert-Butyl-9-oxo-4,8-diazaspiro[4.4]nonane-4-carboxylate

Hazard: Irritant

Formula: C₁₂H₂₀N₂O₃
MW: 240.3 g/mol
MDL Number: MFCD16039601
CAS Number: 1221818-45-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR