tert-Butyl-5-oxo-4,10-diazaspiro[5.5]undecane-10-carboxylate ≥95%

Supplier: Matrix Scientific

069109-250MG 069109-500MG
101938-970EA 1106.15 USD
101938-970 101938-972
tert-Butyl-5-oxo-4,10-diazaspiro[5.5]undecane-10-carboxylate ≥95%
tert-Butyl-5-oxo-4,10-diazaspiro[5.5]undecane-10-carboxylate

Hazard: Irritant

Formula: C₁₄H₂₄N₂O₃
MW: 268.36 g/mol
MDL Number: MFCD08461384
CAS Number: 923009-54-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR