tert-Butyl-4-oxo-3,9-diazaspiro[4.5]decane-9-carboxylate ≥95%

Supplier: Matrix Scientific

Synonyms: tert-Butyl 1-oxo-2,7-diazaspiro[4.5]decane-7-carboxylate

069108-250MG 069108-500MG
101938-966EA 270.77 USD
101938-966 101938-968
tert-Butyl-4-oxo-3,9-diazaspiro[4.5]decane-9-carboxylate ≥95%
tert-Butyl-4-oxo-3,9-diazaspiro[4.5]decane-9-carboxylate

Hazard: Irritant

Formula: C₁₃H₂₂N₂O₃
MW: 254.33 g/mol
MDL Number: MFCD11227060
CAS Number: 923009-50-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR