Methyl-12-oxo-12-phenyldodecanoate ≥95%

Supplier: Matrix Scientific

069107-500MG 069107-250MG
101938-962EA 1106.15 USD
101938-962 101938-964
Methyl-12-oxo-12-phenyldodecanoate ≥95%
Methyl-12-oxo-12-phenyldodecanoate

Hazard: Irritant

Formula: C₁₉H₂₈O₃
MDL Number: MFCD16039591
CAS Number: 104828-35-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR