1-(1-(((9H-Fluoren-9-yl)methoxy)carbonyl)pyrrolidin-3-yl)piperidine-3-carboxylic acid ≥95%

Supplier: Matrix Scientific

069105-250MG 069105-500MG
101938-954EA 901.54 USD
101938-954 101938-956
1-(1-(((9H-Fluoren-9-yl)methoxy)carbonyl)pyrrolidin-3-yl)piperidine-3-carboxylic acid ≥95%
1-(1-(((9H-Fluoren-9-yl)methoxy)carbonyl)pyrrolidin-3-yl)piperidine-3-carboxylic acid

Hazard: Irritant

Formula: C₂₅H₂₈N₂O₄
MDL Number: MFCD19443948
CAS Number: 1313738-82-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR