Dimethyl dodecanedioate ≥95%
Supplier: Matrix Scientific
Synonyms:
Dodecanedioic Acid Dimethyl Ester
069104-25G
069104-5G
069104-1G
101938-950EA
15.38
USD
101938-950
101938-952
101938-948
Dimethyl dodecanedioate ≥95%
Dimethyl dodecanedioate
Purity:
CAS: 1731-79-9
MDL Number: MFCD00043632
Molecular Formula: C14H26O4
Molecular Weight: 258,36
Boiling Point: 187-188/14mm
Hazard: Irritant
Formula:
C₁₄H₂₆O₄ MW: 258.36 g/mol Boiling Pt: 187…188 °C (14 mmHg) Melting Pt: 27…32 °C |
MDL Number:
MFCD00043632 CAS Number: 1731-79-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...