Ethyl-5-Isopropylpyrazole-3-carboxylate ≥98%

Supplier: Matrix Scientific

Synonyms: Ethyl 3-isopropyl-1H-pyrazole-5-carboxylate

069080-1G 069080-5G
101938-840EA 184.62 USD
101938-840 101938-842
Ethyl-5-Isopropylpyrazole-3-carboxylate ≥98%
Ethyl-5-Isopropylpyrazole-3-carboxylate

Hazard: Irritant

Formula: C₉H₁₄N₂O₂
MW: 182.22 g/mol
MDL Number: MFCD08533270
CAS Number: 78208-72-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR