2-Amino-6H-1,3,4-thiadiazine-5-carboxylic acid hydrochloride ≥96%
Supplier: Matrix Scientific
069069-1G
069069-500MG
101938-782EA
815.38
USD
101938-782
101938-784
2-Amino-6H-1,3,4-thiadiazine-5-carboxylic acid hydrochloride ≥96%
2-Amino-6H-1,3,4-thiadiazine-5-carboxylic acid hydrochloride
Hazard: Irritant
Formula:
C₄H₆ClN₃O₂S |
MDL Number:
MFCD06244879 CAS Number: 1171535-57-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...