(R)-Pyrrolidin-3-ylmethanol ≥95%

Supplier: Matrix Scientific

069065-1G 069065-5G
101938-772EA 238.46 USD
101938-772 101938-774
(R)-Pyrrolidin-3-ylmethanol ≥95%
(R)-Pyrrolidin-3-ylmethanol

Hazard: Irritant

Formula: C₅H₁₁NO
MW: 101.15 g/mol
MDL Number: MFCD09607969
CAS Number: 110013-18-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR