2-Aminoisoindoline-1,3-dione hydrochloride ≥95%
Supplier: Matrix Scientific
069050-1G
069050-10G
069050-5G
101938-742EA
204.62
USD
101938-742
101938-744
101938-746
2-Aminoisoindoline-1,3-dione hydrochloride ≥95%
2-Aminoisoindoline-1,3-dione hydrochloride
Hazard: Irritant
Formula:
C₈H₇ClN₂O₂ |
MDL Number:
MFCD13191639 CAS Number: 1198286-64-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...