tert-Butyl-4-(4-methylbenzoyl)piperidine-1-carboxylate ≥95%
Supplier: Matrix Scientific
Synonyms:
tert-Butyl 4-(4-methylbenzoyl)piperidine-1-carboxylate
069046-5G
069046-1G
101938-740EA
633.85
USD
101938-740
101938-738
tert-Butyl-4-(4-methylbenzoyl)piperidine-1-carboxylate ≥95%
tert-Butyl-4-(4-methylbenzoyl)piperidine-1-carboxylate
Hazard: Irritant
Formula:
C₁₈H₂₅NO₃ MW: 303.4 g/mol |
MDL Number:
MFCD08689345 CAS Number: 912768-78-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...