2,2,2-Trifluoro-1-(4-(4-fluorobenzoyl)piperidin-1-yl)ethanone ≥95%
Supplier: Matrix Scientific
069031-250MG
069031-1G
069031-500MG
101938-694EA
263.08
USD
101938-694
101938-696
101938-698
2,2,2-Trifluoro-1-(4-(4-fluorobenzoyl)piperidin-1-yl)ethanone ≥95%
2,2,2-Trifluoro-1-(4-(4-fluorobenzoyl)piperidin-1-yl)ethanone
Hazard: Irritant
Formula:
C₁₄H₁₃F₄NO₂ |
MDL Number:
MFCD11036250 CAS Number: 1159982-32-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...