tert-Butyl-3-benzyl-4-oxo-3,9-diazaspiro[4.5]decane-9-carboxylate ≥95%

Supplier: Matrix Scientific

069026-250MG 069026-500MG
101938-674EA 872.31 USD
101938-674 101938-676
tert-Butyl-3-benzyl-4-oxo-3,9-diazaspiro[4.5]decane-9-carboxylate ≥95%
tert-Butyl-3-benzyl-4-oxo-3,9-diazaspiro[4.5]decane-9-carboxylate

Hazard: Irritant

Formula: C₂₀H₂₈N₂O₃
MW: 344.46 g/mol
MDL Number: MFCD12912609
CAS Number: 1198284-76-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR