tert-Butyl-4-indolin-1-ylpiperidine-1-carboxylate ≥95%
Supplier: Matrix Scientific
069009-1G
069009-500MG
069009-5G
101938-614EA
466.15
USD
101938-614
101938-616
101938-618
tert-Butyl-4-indolin-1-ylpiperidine-1-carboxylate ≥95%
tert-Butyl-4-indolin-1-ylpiperidine-1-carboxylate
Hazard: Irritant
Formula:
C₁₈H₂₆N₂O₂ MW: 302.42 g/mol |
MDL Number:
MFCD12026418 CAS Number: 400828-91-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...