7-(Benzhydryloxy)-1H-indole ≥95%

Supplier: Matrix Scientific

068975-500MG 068975-250MG 068975-1G
101938-498EA 524.62 USD
101938-498 101938-500 101938-502
7-(Benzhydryloxy)-1H-indole ≥95%
7-(Benzhydryloxy)-1H-indole

Hazard: Irritant

Formula: C₂₁H₁₉NO
MDL Number: MFCD07644633
CAS Number: 135328-49-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR