tert-Butyl-3-(2,2,2-trifluoroacetyl)piperidine-1-carboxylate ≥95%
Supplier: Matrix Scientific
068972-250MG
068972-1G
068972-500MG
101938-486EA
769.23
USD
101938-486
101938-488
101938-484
tert-Butyl-3-(2,2,2-trifluoroacetyl)piperidine-1-carboxylate ≥95%
tert-Butyl-3-(2,2,2-trifluoroacetyl)piperidine-1-carboxylate
Hazard: Irritant
Formula:
C₁₂H₁₈F₃NO₃ MW: 281.28 g/mol |
MDL Number:
MFCD11501653 CAS Number: 884512-51-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...