Ethyl-1-benzyl-1,2,5,6-tetrahydropyridine-3-carboxylate ≥95%

Supplier: Matrix Scientific

Synonyms: Ethyl 1-benzyl-1,2,5,6-tetrahydropyridine-3-carboxylate

068971-500MG 068971-250MG 068971-1G
101938-478EA 233.85 USD
101938-478 101938-480 101938-482
Ethyl-1-benzyl-1,2,5,6-tetrahydropyridine-3-carboxylate ≥95%
Ethyl-1-benzyl-1,2,5,6-tetrahydropyridine-3-carboxylate

Hazard: Irritant

Formula: C₁₅H₁₉NO₂
MW: 245.32 g/mol
MDL Number: MFCD01365980
CAS Number: 1159982-41-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR