(6-Chloropyrimidin-4-yl)methanamine ≥95%
Supplier: Matrix Scientific
068969-1G
068969-500MG
068969-250MG
101938-468EA
1396.92
USD
101938-468
101938-470
101938-472
(6-Chloropyrimidin-4-yl)methanamine ≥95%
(6-Chloropyrimidin-4-yl)methanamine
Hazard: Toxic
Formula:
C₅H₆ClN₃ |
MDL Number:
MFCD19982768 CAS Number: 933702-18-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...