1-Benzyl-3-bromo-1,2,5,6-tetrahydropyridine ≥95%

Supplier: Matrix Scientific

068967-500MG 068967-1G
101938-460EA 727.69 USD
101938-460 101938-462
1-Benzyl-3-bromo-1,2,5,6-tetrahydropyridine ≥95%
1-Benzyl-3-bromo-1,2,5,6-tetrahydropyridine

Hazard: Irritant

Formula: C₁₂H₁₄BrN
MDL Number: MFCD12026390
CAS Number: 1159982-62-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR