1-Boc-4-chloropiperidine ≥95%

Supplier: Matrix Scientific

Synonyms: tert-Butyl 4-chloropiperidine-1-carboxylate

068946-5G
101938-386EA 1923.08 USD
101938-386
1-Boc-4-chloropiperidine ≥95%
1-Boc-4-chloropiperidine

Hazard: Irritant

Formula: C₁₀H₁₈ClNO₂
MW: 219.7100 g/mol
MDL Number: MFCD04115040
CAS Number: 154874-94-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR