tert-Butyl-4-(3-nitro-2-pyridyl)piperazine-1-carboxylate ≥95%

Supplier: Matrix Scientific

068945-5G
101938-382EA 2692.31 USD
101938-382
tert-Butyl-4-(3-nitro-2-pyridyl)piperazine-1-carboxylate ≥95%
tert-Butyl-4-(3-nitro-2-pyridyl)piperazine-1-carboxylate

Hazard: Irritant

Formula: C₁₄H₂₀N₄O₄
MW: 308.34 g/mol
MDL Number: MFCD09028514
CAS Number: 153473-24-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR