1-tert-Butyl-4-piperidone ≥95%

Supplier: Matrix Scientific

Synonyms: 1-(tert-Butyl)piperidin-4-one

068944-5G
101938-378EA 1923.08 USD
101938-378
1-tert-Butyl-4-piperidone ≥95%
1-tert-Butyl-4-piperidone

Hazard: Irritant

Formula: C₉H₁₇NO
MW: 155.24 g/mol
MDL Number: MFCD03411608
CAS Number: 1465-76-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR