4-(4-Bromophenyl)-1-tert-butoxycarbonylpiperidine-4-carboxylic acid ≥95%
Supplier: Matrix Scientific
Synonyms:
1-Boc-4-(4-bromophenyl)-4-carboxypiperidine
068939-5G
101938-356EA
1423.08
USD
101938-356
4-(4-Bromophenyl)-1-tert-butoxycarbonylpiperidine-4-carboxylic acid ≥95%
4-(4-Bromophenyl)-1-tert-butoxycarbonylpiperidine-4-carboxylic acid
Hazard: Irritant
Formula:
C₁₇H₂₂BrNO₄ MW: 384.28 g/mol |
MDL Number:
MFCD11226814 CAS Number: 1076197-05-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...