4-Quinazolinecarbonitrile ≥95%

Supplier: Matrix Scientific

068929-500MG
101938-310EA 1458.46 USD
101938-310
4-Quinazolinecarbonitrile ≥95%
4-Quinazolinecarbonitrile

Hazard: Irritant

Formula: C₉H₅N₃
MDL Number: MFCD10697880
CAS Number: 36082-71-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR