5-Amino-1-(2-Fluorobenzyl)-1H-pyrazole ≥95%

Supplier: Matrix Scientific

068927-1G
101938-298EA 440 USD
101938-298
5-Amino-1-(2-Fluorobenzyl)-1H-pyrazole ≥95%
5-Amino-1-(2-Fluorobenzyl)-1H-pyrazole

Hazard: Irritant

Formula: C₁₀H₁₀FN₃
MDL Number: MFCD09817041
CAS Number: 1152858-54-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR