4-Bromo-6-methyl-2-methoxyphenol ≥95%

Supplier: Matrix Scientific

068925-1G
101938-290EA 1318.46 USD
101938-290
4-Bromo-6-methyl-2-methoxyphenol ≥95%
4-Bromo-6-methyl-2-methoxyphenol

Hazard: Irritant

Formula: C₈H₉BrO₂
MDL Number: MFCD09263254
CAS Number: 86232-54-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR