5-Bromoisoindoline ≥95%

Supplier: Matrix Scientific

068922-1G
101938-278EA 384.62 USD
101938-278
5-Bromoisoindoline ≥95%
5-Bromoisoindoline

Hazard: Irritant

Formula: C₈H₈BrN
MDL Number: MFCD08234718
CAS Number: 127168-84-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR