Methyl aminotetrahydropyran-4-ylacetate ≥95%
Supplier: Matrix Scientific
068919-1G
068919-250MG
101938-258EA
524.62
USD
101938-258
101938-262
Methyl aminotetrahydropyran-4-ylacetate ≥95%
Methyl aminotetrahydropyran-4-ylacetate
Hazard: Irritant
Formula:
C₈H₁₅NO₃ |
MDL Number:
MFCD03425227 CAS Number: 477585-43-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...