Nortropine ≥95%

Supplier: Matrix Scientific

Synonyms: (3-endo)-8-Azabicyclo[3.2.1]octan-3-ol8-Azabicyclo[3.2.1]octan-3-ol

068914-25G
101938-234EA 75.38 USD
101938-234
Nortropine ≥95%
Nortropine

Hazard: Irritant

Formula: C₇H₁₃NO
MW: 127.19 g/mol
MDL Number: MFCD00047140
CAS Number: 538-09-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR