3-Aminotetrahydrofuran-3-carboxylic acid ≥95%

Supplier: Matrix Scientific

068908-500MG
101938-200EA 989.23 USD
101938-200
3-Aminotetrahydrofuran-3-carboxylic acid ≥95%
3-Aminotetrahydrofuran-3-carboxylic acid

Hazard: Irritant

Formula: C₅H₉NO₃
MDL Number: MFCD08460976
CAS Number: 125218-55-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR