3-(Dibenzylamino)oxetane-3-carbonitrile ≥95%
Supplier: Matrix Scientific
Synonyms:
3-[Bis(phenylmethyl)amino]-3-oxetanecarbonitrile
068907-1G
068907-250MG
101938-192EA
407.69
USD
101938-192
101938-196
3-(Dibenzylamino)oxetane-3-carbonitrile ≥95%
3-(Dibenzylamino)oxetane-3-carbonitrile
Hazard: Irritant
Formula:
C₁₈H₁₈N₂O MW: 278.36 g/mol |
MDL Number:
MFCD18432775 CAS Number: 1021393-00-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...