5-Amino-7-fluoro-1H-indole ≥95%

Supplier: Matrix Scientific

068905-500MG
101938-184EA 2061.54 USD
101938-184
5-Amino-7-fluoro-1H-indole ≥95%
5-Amino-7-fluoro-1H-indole

Hazard: Irritant

Formula: C₈H₇FN₂
MDL Number: MFCD12923890
CAS Number: 926028-84-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR