4-Ethyl-5-fluoropyrimidine ≥95%

Supplier: Matrix Scientific

068889-1G
101938-108EA 40 USD
101938-108
4-Ethyl-5-fluoropyrimidine ≥95%
4-Ethyl-5-fluoropyrimidine

Hazard: Irritant

Formula: C₆H₇FN₂
MDL Number: MFCD06657566
CAS Number: 137234-88-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR