[(1, 1-Dioxotetrahydro-2H-thiopyran-4-yl)methyl]amine ≥95%

Supplier: Matrix Scientific

068885-500MG
101938-090EA 1209.23 USD
101938-090
[(1, 1-Dioxotetrahydro-2H-thiopyran-4-yl)methyl]amine ≥95%
[(1, 1-Dioxotetrahydro-2H-thiopyran-4-yl)methyl]amine

Hazard: Irritant

Formula: C₆H₁₃NO₂S
MDL Number: MFCD09864365
CAS Number: 476660-77-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR