1H-1,2,4-Triazole-3-sulfonamide ≥95%

Supplier: Matrix Scientific

068884-1G
101938-086EA 1758.46 USD
101938-086
1H-1,2,4-Triazole-3-sulfonamide ≥95%
1H-1,2,4-Triazole-3-sulfonamide

Hazard: Irritant

Formula: C₂H₄N₄O₂S
MDL Number: MFCD18207305
CAS Number: 89517-96-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR