trans-tert-Butyl-N-(3-aminocyclobutyl)carbamate ≥95%

Synonyms: trans-Cyclobutane-1,3-diamine, N1-BOC protectedtert-Butyl (trans-3-aminocyclobutyl)carbamate

068882-250MG
101938-072EA 243.62 USD
101938-072
trans-tert-Butyl-N-(3-aminocyclobutyl)carbamate ≥95%
trans-tert-Butyl-N-(3-aminocyclobutyl)carbamate

Hazard: Irritant

Formula: C₉H₁₈N₂O₂
MW: 186.26 g/mol
MDL Number: MFCD09040641
CAS Number: 871014-19-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR