1-(5-Fluoropyridin-2-yl)ethanamine ≥95%

Supplier: Matrix Scientific

068875-250MG
101938-044EA 300 USD
101938-044
1-(5-Fluoropyridin-2-yl)ethanamine ≥95%
1-(5-Fluoropyridin-2-yl)ethanamine

Hazard: Irritant

Formula: C₇H₉FN₂
MDL Number: MFCD18260255
CAS Number: 915720-57-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR