4-Phenyl-5,6-dihydro-2H-pyran ≥95%

Supplier: Matrix Scientific

068870-500MG
101938-020EA 1309.23 USD
101938-020
4-Phenyl-5,6-dihydro-2H-pyran ≥95%
4-Phenyl-5,6-dihydro-2H-pyran

Hazard: Irritant

Formula: C₁₁H₁₂O
MDL Number: MFCD18432773
CAS Number: 3174-81-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR